Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-67
Entrez Gene ID
184639
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F18E3.2
Chromosome Location
Chr.V: 7417660 - 7419828 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072499.3 NP_504900.2 4 - 5 809 107

View Full Product Details