Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-28
Entrez Gene ID
191971
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F25E5.12
Chromosome Location
Chr.V: 7471651 - 7472915 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072520.1 NP_504921.1 3 - 4 544 155

View Full Product Details