Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-8
Entrez Gene ID
182772
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C18B10.5
Chromosome Location
Chr.V: 7483332 - 7485100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072524.5 NP_504925.2 2 - 3 343 67

View Full Product Details