Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srb-19
Entrez Gene ID
183809
Gene Name
Serpentine Receptor, class B (beta)
Gene Aliases
CELE_C54F6.11
Chromosome Location
Chr.V: 7517652 - 7520177 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072535.3 NP_504936.2 1 - 2 418 186

View Full Product Details