Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33A1
Entrez Gene ID
179148
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C12D5.7
Chromosome Location
Chr.V: 7679314 - 7681615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072587.5 NP_504988.1 2 - 3 682 74

View Full Product Details