Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-9
Entrez Gene ID
188110
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_T05B11.2
Chromosome Location
Chr.V: 7738010 - 7739596 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072599.2 NP_505000.2 4 - 5 799 116

View Full Product Details