Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptl-2
Entrez Gene ID
266646
Gene Name
protein-coding
Gene Aliases
CELE_F43H9.2
Chromosome Location
Chr.V: 8016665 - 8021715 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072663.6 NP_505064.1 4 - 5 533 60
NM_072664.5 NP_505065.1 3 - 4 426 60

View Full Product Details