Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-90
Entrez Gene ID
191995
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F58G4.2
Chromosome Location
Chr.V: 8103260 - 8107022 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072691.3 NP_505092.1 5 - 6 563 145

View Full Product Details