Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
myo-5
Entrez Gene ID
179190
Gene Name
MYOsin heavy chain structural genes
Gene Aliases
CELE_F58G4.1
Chromosome Location
Chr.V: 8108970 - 8118131 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072693.4 NP_505094.1 1 - 2 139 92

View Full Product Details