Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sul-2
Entrez Gene ID
179194
Gene Name
Sulfatase domain-containing protein
Gene Aliases
CELE_D1014.1
Chromosome Location
Chr.V: 8157746 - 8159872 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072701.6 NP_505102.1 2 - 3 1006 139

View Full Product Details