Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pax-1
Entrez Gene ID
187105
Gene Name
Paired domain-containing protein
Gene Aliases
CELE_K07C11.1
Chromosome Location
Chr.V: 8224934 - 8226116 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072719.2 NP_505120.1 1 - 2 190 93

View Full Product Details