Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-33
Entrez Gene ID
184786
Gene Name
Serpentine receptor class gamma-33
Gene Aliases
CELE_F21F8.1
Chromosome Location
Chr.V: 8292064 - 8293516 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072739.1 NP_505140.1 5 - 6 840 135

View Full Product Details