The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_072756.5 | NP_505157.1 | 1 - 2 | 125 | 63 |
GenBank mRNA | AY008125.1 | - | 1 - 2 | 116 | 63 |