Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsp-17
Entrez Gene ID
186113
Gene Name
protein-coding
Gene Aliases
CELE_F52E1.7
Chromosome Location
Chr.V: 8384799 - 8385933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028786.7 NP_001023957.1 1 - 2 63 63

View Full Product Details