Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-7
Entrez Gene ID
179222
Gene Name
protein-coding
Gene Aliases
CELE_F52E1.4
Chromosome Location
Chr.V: 8387356 - 8392324 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028783.1 NP_001023954.1 11 - 12 1615 81
NM_001028784.1 NP_001023955.1 11 - 12 1615 81
NM_001373061.1 NP_001360539.1 11 - 12 1615 81

View Full Product Details