Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pxn-1
Entrez Gene ID
191484
Gene Name
Peroxidasin homolog pxn-1
Gene Aliases
CELE_ZK994.3
Chromosome Location
Chr.V: 8494889 - 8502442 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072787.4 NP_505188.3 19 - 20 3557 81

View Full Product Details