Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-6
Entrez Gene ID
185807
Gene Name
OToPetrin-Like
Gene Aliases
CELE_F45F2.6
Chromosome Location
Chr.V: 8525908 - 8528580 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072794.2 NP_505195.1 5 - 6 1199 147

View Full Product Details