Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-5
Entrez Gene ID
185806
Gene Name
OToPetrin-Like
Gene Aliases
CELE_F45F2.5
Chromosome Location
Chr.V: 8529519 - 8532465 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072795.4 NP_505196.2 3 - 4 553 64

View Full Product Details