Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-4
Entrez Gene ID
185805
Gene Name
OToPetrin-Like
Gene Aliases
CELE_F45F2.1
Chromosome Location
Chr.V: 8537331 - 8539614 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072801.2 NP_505202.2 4 - 5 624 124

View Full Product Details