Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-15
Entrez Gene ID
191602
Gene Name
AcetylCholine Receptor
Gene Aliases
CELE_F25G6.4
Chromosome Location
Chr.V: 8552695 - 8555388 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072805.5 NP_505206.2 5 - 6 803 69

View Full Product Details