Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smk-1
Entrez Gene ID
179243
Gene Name
protein-coding
Gene Aliases
CELE_F41E6.4
Chromosome Location
Chr.V: 8607800 - 8615841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028738.7 NP_001023909.2 3 - 4 676 71
NR_138495.1 3 - 4 674 71

View Full Product Details