Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-87
Entrez Gene ID
191994
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F59E11.16
Chromosome Location
Chr.V: 8986574 - 8988204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072921.2 NP_505322.1 6 - 7 1185 100

View Full Product Details