The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_072940.5 | NP_505341.1 | 9 - 10 | 1277 | 132 |
NR_145827.1 | 7 - 8 | 1011 | 132 |