Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33B1
Entrez Gene ID
182888
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C25E10.2
Chromosome Location
Chr.V: 9041330 - 9043677 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072941.2 NP_505342.2 6 - 7 980 93

View Full Product Details