Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fip-2
Entrez Gene ID
179283
Gene Name
Fungus-Induced Protein
Gene Aliases
CELE_T27E4.4
Chromosome Location
Chr.V: 9067976 - 9068390 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072949.6 NP_505350.1 1 - 2 213 74

View Full Product Details