Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spl-2
Entrez Gene ID
181859
Gene Name
Sphingosine Phosphate Lyase
Gene Aliases
CELE_B0222.4
Chromosome Location
Chr.V: 9144616 - 9146899 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072971.6 NP_505372.1 3 - 4 672 77

View Full Product Details