Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-10
Entrez Gene ID
188580
Gene Name
protein-coding
Gene Aliases
CELE_T18H9.5
Chromosome Location
Chr.V: 9204661 - 9207477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028968.4 NP_001024139.1 1 - 2 215 143
NM_001028969.6 NP_001024140.1 1 - 2 221 143
NM_001373070.2 NP_001360570.1 2 - 2 225 143

View Full Product Details