Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mdt-27
Entrez Gene ID
179303
Gene Name
MeDiaTor
Gene Aliases
CELE_T18H9.6
Chromosome Location
Chr.V: 9209293 - 9213654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072985.7 NP_505386.2 6 - 7 1153 90

View Full Product Details