Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aat-2
Entrez Gene ID
184126
Gene Name
Amino Acid Transporter
Gene Aliases
CELE_F07C3.7
Chromosome Location
Chr.V: 9244402 - 9246360 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072993.6 NP_505394.2 1 - 2 279 135

View Full Product Details