Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-94
Entrez Gene ID
191996
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F07C3.8
Chromosome Location
Chr.V: 9248031 - 9249306 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072994.2 NP_505395.2 3 - 4 469 142

View Full Product Details