Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-10
Entrez Gene ID
191402
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_ZK682.4
Chromosome Location
Chr.V: 9280512 - 9282210 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073000.4 NP_505401.2 3 - 4 648 150

View Full Product Details