Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-39
Entrez Gene ID
185720
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_F44A2.4
Chromosome Location
Chr.V: 9303326 - 9304622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073004.1 NP_505405.1 2 - 3 428 152

View Full Product Details