Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acl-14
Entrez Gene ID
179313
Gene Name
protein-coding
Gene Aliases
CELE_K07B1.5
Chromosome Location
Chr.V: 9327141 - 9333631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383368.1 5 - 6 935 78
NM_073011.8 NP_505412.2 5 - 6 972 78

View Full Product Details