Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-5
Entrez Gene ID
187088
Gene Name
Trypsin-like protease try-5
Gene Aliases
CELE_K07B1.1
Chromosome Location
Chr.V: 9345772 - 9348687 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073020.5 NP_505421.3 6 - 7 925 92
GenBank mRNA JN651275.1 - 6 - 7 925 92
JN651276.1 - 6 - 7 956 92
JN651277.1 - 5 - 6 834 92

View Full Product Details