Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acs-14
Entrez Gene ID
179330
Gene Name
fatty Acid CoA Synthetase family
Gene Aliases
CELE_F11A3.1
Chromosome Location
Chr.V: 9508132 - 9510435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073050.6 NP_505451.1 6 - 7 1514 85

View Full Product Details