Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dop-2
Entrez Gene ID
179347
Gene Name
protein-coding
Gene Aliases
CELE_K09G1.4
Chromosome Location
Chr.V: 9608556 - 9614942 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028876.3 NP_001024047.1 8 - 9 930 122
NM_001269197.4 NP_001256126.1 9 - 10 1368 122
NM_001269198.4 NP_001256127.1 9 - 10 1018 122
NM_001373076.2 NP_001360549.1 9 - 9 1344 122
NM_001373077.2 NP_001360548.1 9 - 9 999 122
NM_001373078.2 NP_001360550.1 8 - 9 928 122
GenBank mRNA AB125702.1 - 8 - 9 952 122

View Full Product Details