Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-63
Entrez Gene ID
188181
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_T06E4.11
Chromosome Location
Chr.V: 9616031 - 9617651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073078.6 NP_505479.3 2 - 2 1373 86

View Full Product Details