Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-29
Entrez Gene ID
183645
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C50C10.2
Chromosome Location
Chr.V: 9811454 - 9812803 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073131.2 NP_505532.1 3 - 4 806 83

View Full Product Details