Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-9
Entrez Gene ID
179382
Gene Name
protein-coding
Gene Aliases
CELE_C50H2.3
Chromosome Location
Chr.V: 9907615 - 9915121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269224.3 NP_001256153.1 1 - 2 141 123

View Full Product Details