Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-33
Entrez Gene ID
188213
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T07C12.1
Chromosome Location
Chr.V: 9935548 - 9937535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073156.1 NP_505557.1 4 - 5 660 104

View Full Product Details