Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-41
Entrez Gene ID
179387
Gene Name
protein-coding
Gene Aliases
CELE_C27H6.1
Chromosome Location
Chr.V: 9966380 - 9976238 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269228.3 NP_001256157.1 9 - 10 4178 130
NM_001269229.3 NP_001256158.1 7 - 8 3320 130
NM_001269230.3 NP_001256159.1 5 - 6 3047 130

View Full Product Details