Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uda-1
Entrez Gene ID
179395
Gene Name
Nucleoside-diphosphatase uda-1
Gene Aliases
CELE_K08H10.4
Chromosome Location
Chr.V: 10004755 - 10007482 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073172.4 NP_505573.1 4 - 5 1111 82

View Full Product Details