Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-47
Entrez Gene ID
187570
Gene Name
Putative UDP-glucuronosyltransferase ugt-47
Gene Aliases
CELE_R04B5.9
Chromosome Location
Chr.V: 10096305 - 10098597 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073194.5 NP_505595.2 1 - 2 174 112

View Full Product Details