Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-27
Entrez Gene ID
183704
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C51E3.2
Chromosome Location
Chr.V: 10150028 - 10152248 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073208.5 NP_505609.2 2 - 3 573 151

View Full Product Details