Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-3
Entrez Gene ID
179412
Gene Name
protein-coding
Gene Aliases
CELE_C51E3.7
Chromosome Location
Chr.V: 10169177 - 10173753 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028561.4 NP_001023732.1 5 - 6 1053 90
NM_001269263.3 NP_001256192.1 4 - 5 676 90
GenBank mRNA U04995.1 - 5 - 6 1073 90

View Full Product Details