Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul-5
Entrez Gene ID
179413
Gene Name
Cullin-5
Gene Aliases
CELE_ZK856.1
Chromosome Location
Chr.V: 10178977 - 10183249 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073215.7 NP_505616.2 10 - 11 1573 107

View Full Product Details