Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-36
Entrez Gene ID
179422
Gene Name
Cholesterol 7-desaturase
Gene Aliases
CELE_C12D8.5
Chromosome Location
Chr.V: 10223393 - 10226206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073228.6 NP_505629.2 2 - 3 255 70

View Full Product Details