Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- set-22
- Entrez Gene ID
-
179457
- Gene Name
- SET domain-containing protein
- Gene Aliases
-
CELE_Y32F6A.1
- Chromosome Location
-
Chr.V: 10437003 - 10441159 on Build WBcel235
- Gene Symbol
- pap-1
- Entrez Gene ID
-
179458
- Gene Name
- Poly(A) polymerase
- Gene Aliases
-
CELE_Y32F6A.3
- Chromosome Location
-
Chr.V: 10436959 - 10446472 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383385.1 |
|
3 - 4 |
364 |
69 |
View Full Product Details