Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- aqp-6
- Entrez Gene ID
-
183121
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C32C4.2
- Chromosome Location
-
Chr.V: 10648446 - 10651997 on Build WBcel235
- Gene Symbol
- C32C4.16
- Entrez Gene ID
-
13214870
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_C32C4.16
- Chromosome Location
-
Chr.V: 10647561 - 10651957 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383387.1 |
|
6 - 7 |
1621 |
142 |
View Full Product Details