Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqv-4
Entrez Gene ID
179484
Gene Name
UDP-glucose 6-dehydrogenase
Gene Aliases
CELE_F29F11.1
Chromosome Location
Chr.V: 10659599 - 10661467 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073329.10 NP_505730.1 3 - 4 1055 113
GenBank mRNA AY147932.1 - 3 - 4 1057 113

View Full Product Details